missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-DIPAMP, 97%

Product Code. 15423817
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423817 250 mg 250mg
11357569 100 mg 100mg
11377569 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15423817 Supplier Thermo Scientific Alfa Aesar Supplier No. H28037.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 97858-62-3 | C28H28O2P2 | 458.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97858-62-3
Molecular Formula C28H28O2P2
Molecular Weight (g/mol) 458.48
MDL Number MFCD05863547
InChI Key QKZWXPLBVCKXNQ-UHFFFAOYNA-N
Synonym 1,2-bis s-2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp, unii-hs6f5ew3u6, hs6f5ew3u6, ethylenebis s-o-anisyl phenyl phosphine, s,s-+-1,2-bis 2-methoxyphenyl phenyl phosphino ethane, s,s-dipamp; s,s-ethylenebis 2-methoxyphenyl phenylphosphine, s-2-methoxyphenyl 2-s-2-methoxyphenyl phenyl phosphanyl ethyl phenylphosphane
PubChem CID 11419748
IUPAC Name (S)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane
SMILES COC1=CC=CC=C1P(CCP(C1=CC=CC=C1)C1=CC=CC=C1OC)C1=CC=CC=C1

Specifications

Melting Point 101°C to 105°C
Quantity 250 mg
Optical Rotation +84° (c=1 in Chloroform)
Formula Weight 458.47
Percent Purity 97%
Chemical Name or Material (S,S)-DIPAMP

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.