missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-Palonosetron Hydrochloride, TRC

Product Code. 30425705
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425705 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425705 Supplier Toronto Research Chemicals Supplier No. TRCP165800250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Palonosetron Hydrochloride
CAS 135729-62-3
Molecular Formula C19 H24 N2 O . Cl H
Quantity 250 mg
Synonym 1H-Benz[de]isoquinolin-1-one','2-[(3S)-1-azabicyclo[2.2.2]oct-3-yl]-2,3,3a,4,5,6-hexahydro-','hydrochloride (1:1)','(3aS)-, 1H-Benz[de]isoquinolin-1-one','2-(1-azabicyclo[2.2.2]oct-3-yl)-2,3,3a,4,5,6-hexahydro-','monohydrochloride','[S-(R*,R*)]-, 1H-Benz[de]isoquinolin-1-one','2-(3S)-1-azabicyclo[2.2.2]oct-3-yl-2,3,3a,4,5,6-hexahydro-','monohydrochloride','(3aS)- (9CI), Aloxi, Onicit, Palonosetron hydrochloride, RS 25259-197
InChI Formula InChI=1S/C19H24N2O.ClH/c22-19-16-6-2-4-14-3-1-5-15(18(14)16)11-21(19)17-12-20-9-7-13(17)8-10-20;/h2,4,6,13,15,17H,1,3,5,7-12H2;1H/t15-,17-;/m1./s1
SMILES Cl.O=C1N(C[C@H]2CCCc3cccc1c23)[C@@H]4CN5CCC4CC5
IUPAC Name (3aS)-2-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-3a,4,5,6-tetrahydro-3H-benzo[de]isoquinolin-1-one;hydrochloride
Molecular Weight (g/mol) 332.87
Formula Weight 332.1655
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.