missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ S-Sulfo-L-cysteine sodium salt

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
S-Sulfo-L-cysteine sodium salt is an EAA receptor agonist. Agonist at mGlu1α and mGlu5a subtypes expressed in clonal RGT cell lines.
Specifications
Specifications
| CAS | 7381-67-1 |
| Quantity | 50 mg |
| Solubility | Soluble to 100 mM in water |
| Molecular Formula | C3H6NO5S2Na |
| Formula Weight | Observed MW: 223.19 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction