missing translation for 'onlineSavingsMsg'
Learn More

Solifenacin hydrochloride, MedChemExpress

Product Code. 16418808
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16418808

Brand: Medchem Express HYI0230

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Solifenacin hydrochloride (YM905 hydrochloride) is a muscarinic receptor antagonist, with pKis of 7.6, 6.9 and 8.0 for M1, M2 and M3 receptors, respectively.

Solifenacin hydrochloride (YM905 hydrochloride) is a muscarinic receptor antagonist, with pKis of 7.6, 6.9 and 8.0 for M1, M2 and M3 receptors, respectively.

Specifications

Chemical Name or Material Solifenacin hydrochloride
CAS 180468-39-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H27ClN2O2
Quantity 10 mg
Synonym YM905 hydrochloride
Solubility Information DMSO : 50 mg/mL (125.34 mM; Need ultrasonic) ∣H2O : 50 mg/mL (125.34 mM; Need ultrasonic)
SMILES O=C(O[C@H]1CN2CCC1CC2)N3[C@H](C4=C(CC3)C=CC=C4)C5=CC=CC=C5.Cl
Molecular Weight (g/mol) 398.93
Formula Weight 398.93
Percent Purity 99.29%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.