missing translation for 'onlineSavingsMsg'
Learn More

S-tert-Butyl-L-cysteine hydrochloride, 98%

Product Code. 15495488
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495488 25 g 25g
15485488 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495488 Supplier Thermo Scientific Alfa Aesar Supplier No. H59244.14
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2481-09-6 | C7H16ClNO2S | 213.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2481-09-6
MDL Number MFCD00037426

Specifications

Color White
Molecular Formula C7H16ClNO2S
Synonym s-tert-butyl-l-cysteine hydrochloride, h-cys tbu-oh.hcl, h-cys tbu-oh hcl, s-t-butyl-l-cysteine hydrochloride, h-cys tbu-ohhcl, s-tert-butyl-l-cysteine-hydrogen chloride 1/1, 2r-2-amino-3-tert-butylsulfanyl propanoic acid hydrochloride, pubchem19026, h-cys but-oh hcl, h-cys but-oh hydrochloride
InChI Key MHBMYFJKEBCMDR-JEDNCBNOSA-N
SMILES CC(C)(C)SCC(C(=O)O)N.Cl
IUPAC Name (2R)-2-amino-3-tert-butylsulfanylpropanoic acid;hydrochloride
Molecular Weight (g/mol) 213.72
Quantity 25 g
PubChem CID 17750295
Formula Weight 213.73
Percent Purity 98%
Physical Form Powder
Chemical Name or Material S-tert-Butyl-L-cysteine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.