missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Tetrahydro-2-furoic acid, 98+%, ee 98%, Thermo Scientific™

Product Code. 11388706
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388706 1 g 1g
11398706 5 g 5g
11308716 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388706 Supplier Thermo Scientific Alfa Aesar Supplier No. L19411.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87392-07-2
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00272639
InChI Key UJJLJRQIPMGXEZ-BYPYZUCNSA-N
Synonym s-tetrahydrofuran-2-carboxylic acid, s---2-tetrahydrofuroic acid, s---tetrahydro-2-furoic acid, 2s-oxolane-2-carboxylic acid, s-tetrahydro-2-furoic acid, s---tetrahydrofuran-2-carboxylic acid, unii-0i2b46k2sj, tetrahydro-2-furoic acid,-, 2-furancarboxylic acid, tetrahydro-, 2s, s---2-carboxytetrahydrofuroic acid
PubChem CID 446666
ChEBI CHEBI:45960
IUPAC Name (2S)-oxolane-2-carboxylic acid
SMILES C1CC(OC1)C(=O)O

Specifications

Density 1.213
Quantity 1 g
Refractive Index 1.46
UN Number UN3265
Beilstein 4658738
Boiling Point 244°C
Formula Weight 116.12
Percent Purity ≥98%
Assay ee 98%
Chemical Name or Material (S)-(-)-Tetrahydro-2-furoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.