missing translation for 'onlineSavingsMsg'
Learn More

s-Triazine-1,3,5-triethanol, TRC

Product Code. 30136122
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136122 10 g 10g
30134636 1 g 1g
18071410 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136122 Supplier Toronto Research Chemicals Supplier No. TRCT76734510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4719-04-4
Formula Weight 219.1583
InChI Formula InChI=1S/C9H21N3O3/c13-4-1-10-7-11(2-5-14)9-12(8-10)3-6-15/h13-15H,1-9H2
IUPAC Name 2-[3,5-bis(2-hydroxyethyl)-1,3,5-triazinan-1-yl]ethanol
Molecular Formula C9 H21 N3 O3
Molecular Weight (g/mol) 219.28
SMILES OCCN1CN(CCO)CN(CCO)C1
Synonym s-Triazine-1,3,5(2H,4H,6H)-triethanol (8CI), 1,3,5-Tris(2-hydroxyethyl)-1,3,5-triazacyclohexane, 1,3,5-Tris(2-hydroxyethyl)hexahydro-1,3,5-triazine, 1,3,5-Tris(2-hydroxyethyl)hexahydro-s-triazine, Actane, Acticide GR, BK, Bactraclean, Bestcide 1087T, Bioban GK, Busan 1060, Busan 1506, Cobate C, Dodigen NJ, ETA 75, Grotan B, Grotan BK, Hexahydro-1,3,5-tri(2-hydroxyethyl)-s-triazine, Hexahydro-1,3,5-tris(2-hydroxyethyl)-s-triazine, Hexahydro-1,3,5-tris(hydroxyethyl)-s-triazine, KM 200, KM 200 (alcohol), Kalpur TE, Mergal 165, N,N',N' '-Tris(2-hydroxyethyl)hexahydro-s-triazine, N,N',N' '-Tris(β-hydroxyethyl)hexahydro-s-triazine, NSC 516387, Nipacide BK, Onyxide 200, Permachem OB 2, Protectol HT, Roksol T 1-7, Surcide D, Surcide P, Triadine 174, Triadine 3
Chemical Name or Material s-Triazine-1,3,5-triethanol

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.