missing translation for 'onlineSavingsMsg'
Learn More

s-Trioxane, 99+%

Product Code. 10021610 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10021610 2 kg Plastic bottle 2kg
10407160 5 g Glass bottle 5g
10227313 100 g Plastic bottle 100g
10636152 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10021610 Supplier Thermo Scientific Acros Supplier No. 140290020
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 110-88-3 | C3H6O3 | 90.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-88-3
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00006563
InChI Key BGJSXRVXTHVRSN-UHFFFAOYSA-N
Synonym s-trioxane, trioxymethylene, metaformaldehyde, triformol, formaldehyde, trimer, aldeform, formagene, marvosan, trioxan, sym-trioxane
PubChem CID 8081
ChEBI CHEBI:38043
IUPAC Name 1,3,5-trioxane
SMILES C1OCOCO1

Specifications

Melting Point 61.0°C to 62.0°C
Color Colorless to White
Density 1.1700g/mL
Boiling Point 114.0°C to 116.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Quantity 2 kg
Beilstein 19,381
Merck Index 15,9906
Specific Gravity 1.17
Solubility Information Solubility in water: soluble in water: 221g/L (25°C). Other solubilities: soluble in alcohols,ether,ketones,acetone,,chlorinatid and aromatic hydrocarbons,and,other organic solvents,slightly soluble in pentane and petr.ether
Sublimation Point 0115.0°C
Formula Weight 90.08
Percent Purity 99+%
Physical Form Crystalline Flakes or Crystals
Chemical Name or Material s-Trioxane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.