Learn More
Thermo Scientific™ S1 Pipet Filler Accessories
Product Code. 10193923
Change view
Click to view available options
Type:
Nosepiece
Pipette Filter, Non-sterile
Pipette Filter, Sterile
Silicone Pipet Gripper
Table Mount Stand
Universal Power Supply
Wall Mount Stand
Wing Stand
Unit Size:
Each
Pack of 25
Pack of 4
Pack of 5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10193923 Pipette Filter, Sterile Pack of 25
10339352 Pipette Filter, Non-sterile Pack of 5
10030524 Nosepiece Each
10200124 Silicone Pipet Gripper Pack of 4
10375474 Wall Mount Stand Each
10335524 Table Mount Stand Each
11866244 Universal Power Supply Each
11992181 Wing Stand Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10193923 Supplier Thermo Scientific™ Supplier No. 9057
Spotlight Offers
Use promo code 28037
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
Incorporate Thermo Scientific™ S1 Pipet Filler Accessories with S1 Pipet Filler.
Complete your S1 Pipet Filler with these accessories and prolong its use.
Features:
  • Wall-mounted holder and table stand offer compact storage between applications. Recharge the pipet filler in both stands by plugging in the charger. Store pipet filler when working in laminar flow cabinets to save space.
  • Wing stand allows user to set the pipet filler down without removing or emptying the serological pipet.
  • Easy to change filter prevents aerosol or liquid cross contamination of the pipet together with autoclavable silicone adapter.
  • 1mL Pipet Support ensures that 1 mL pipets will hold as securely as larger sizes.
Note:
Wing stand not available for use or sale within the United States or for importation into the United States.
TRUSTED_SUSTAINABILITY
Specifications
Description Hydrophobic Filters, 0.45µm, Sterile
For Use With S1 Pipette Filler
Type Pipette Filter, Sterile