missing translation for 'onlineSavingsMsg'
Learn More

Saccharin, Extra Pure, SLR, Fisher Chemical

Product Code. 10645502 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10645502 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10645502 Supplier Fisher Chemical Supplier No. S/0040/48
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 81-07-2
Melting Point 226°C
Color White
pH 2
Packaging Plastic powder jar
Molecular Formula C7H5NO3S
MDL Number 5866
Quantity 100 g
Synonym saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinose
InChI Key CVHZOJJKTDOEJC-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=O)NS2(=O)=O
IUPAC Name 1,1-dioxo-1,2-benzothiazol-3-one
Molecular Weight (g/mol) 183.181
PubChem CID 5143
ChEBI CHEBI:32111
Formula Weight 183.18g/mol
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.