missing translation for 'onlineSavingsMsg'
Learn More

Saccharin, sodium salt hydrate, 99+%

Product Code. 10722271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10722271 250 g Plastic bottle 250g
10553391 1 kg Plastic bottle 1kg
10145113 5 kg Plastic Drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10722271 Supplier Thermo Scientific Acros Supplier No. 223372500
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 82385-42-0 | C7H4NNaO3S | 205.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82385-42-0
Molecular Formula C7H4NNaO3S
Molecular Weight (g/mol) 205.16
MDL Number MFCD00149605
InChI Key OAZGZMKGRPRHJX-UHFFFAOYSA-M
Synonym saccharin sodium salt hydrate
PubChem CID 131673955
IUPAC Name 1,1-dioxo-1,2-benzothiazol-3-one;molecular hydrogen;sodium;hydrate
SMILES [Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12

Specifications

Color Colorless to White
Infrared Spectrum Authentic
Assay Percent Range 25ppm Toluenesulfonamides (GC) max
Packaging Plastic bottle
Quantity 250 g
Beilstein 27,17
Merck Index 10,8171
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol, practically insoluble in diethyl ether and, chloroform
Formula Weight 205.16
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Saccharin, sodium salt hydrate, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.