missing translation for 'onlineSavingsMsg'
Learn More

Saccharin, TRC

Product Code. 30012671
Change view
Click to view available options
Quantity:
100 mg
1 g
50 g
Unit Size:
100mg
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012671 100 mg 100mg
30005460 50 g 50g
30005542 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012671 Supplier Toronto Research Chemicals Supplier No. TRCS080800100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-07-2
Molecular Formula C7 H5 N O3 S
Molecular Weight (g/mol) 183.18
Synonym Saccharin, 1,2-Benzisothiazol-3(2H)-one 1,1-dioxide, 1,2-Benzisothiazolin-3-one, 1,1-dioxide (8CI), 1,1-Dioxido-3-oxo-2,3-dihydrobenzo[d]isothiazole, 1,1-Dioxo-1,2-benzisothiazol-3(2H)-one, 1,1-Dioxo-1,2-benzothiazol-3-one, 1,2-Dihydro-2-ketobenzisosulfonazole, 2,3-Dihydro-1,2-benzisothiazol-3-one-1,1-dioxide, 2,3-Dihydro-3-oxobenzisosulfonazole, 3-Benzisothiazolinone 1,1-dioxide, 3-Hydroxybenzisothiazole-S,S-dioxide, 550 Saccharine, Anhydro-o-sulfaminebenzoic acid, Benzoic sulfimide, Benzoic sulphinide, Benzosulfimide, Benzosulfinide, Garantose, Glucid, Gluside, NSC 5349, Necta Sweet, Saccharimide, Saccharin, Saccharin acid, Saccharin insoluble, Saccharine, Saccharinol, Saccharinose, Saccharol, o-Benzoic acid sulfimide, o-Benzoic sulfimide, o-Benzosulfimide, o-Benzoyl sulfimide, o-Sulfobenzimide, o-Sulfobenzoic acid imide
IUPAC Name 1,1-dioxo-1,2-benzothiazol-3-one
SMILES O=C1NS(=O)(=O)c2ccccc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H5NO3S/c9-7-5-3-1-2-4-6(5)12(10,11)8-7/h1-4H,(H,8,9)
Formula Weight 182.999
Percent Purity >95%
Chemical Name or Material Saccharin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.