missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ Safe Aspiration Station

Product Code. 17193574 Shop All Gilson Products
Change view
Click to view available options
Plug Type:
EU
UK
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Unit Size
17193574 UK Each
17102225 EU Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17193574 Supplier Gilson™ Supplier No. F110746
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Safe Aspiration Station is a stand-alone motorized vacuum system for quick and easy aspiration of biological liquid waste disposal. The station offers adjustable aspiration speed and handles. Ideal for ELISA tests, cell culture, and any other contaminated or hazardous liquid aspiration and disposal.

Safe and Contamination-Free

  • Closed, stand-alone design to prevent contamination hazards
  • All parts in contact with liquids are fully autoclavable
  • Equipped with a level detection system to prevent liquid overflow
  • Features an in-line hydrophobic filter that traps aerosols for safe disposal

Adaptable

  • Various adapters included to be fitted to the hand operator
  • Adjustable aspiration level indicated by a board of LEDs
  • Modular design enables quick and accurate aspiration of liquids

Convenient

  • Easy to use on a bench or in laminar flow cabinets
  • Always ready to use by simply pressing the button on the hand operator
  • Ergonomic hand operator can be stored on the bottle handle
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Blue
Dispensing Speed 17 mL/sec.
Disposable Reusable
Grip Type Ergonomic
Material Polypropylene
Number of Channels 1
Product Line MLH Product Line
Sterility Non-sterile
Type Motorized Vacuum
Volume (Metric) 4 L
Plug Type UK
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.