Learn More
Hirschmann™ Safe-Cone Filters for Labopette™
For the prevention of cross contamination of liquid samples or vapours when pipetting radioactive, biohazardous or corrosive samples.
Brand:  Hirschmann™ 9470013
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10012223
  • 70.55 € / Pack of 50
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
  • Provides the decisive advantage when protecting pipettes and samples against contamination
  • Extends the service life of pipettes
  • Provides excellent protection for the internal components if replaced regularly
  • Can serve as additional protection against penetration of liquids into the interior of the pipette
  • For use with a range of Labopette™ models
Specifications
Specifications
Safety filter
Protection against cross-contamination
Labopette models 9475406, 9475407, 9475503, 9475504, 9475505, 9475506, 9475602, 9475702