Learn More
Hirschmann™ Safe-Cone Filters for Labopette™
For the prevention of cross contamination of liquid samples or vapours when pipetting radioactive, biohazardous or corrosive samples.
70.20 € - 70.55 €
Specifications
Type | Safety filter |
---|---|
For Use With (Application) | Protection against cross-contamination |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10233151
|
Hirschmann™
9470010 |
Labopette models 9475404, 9475405, 9475511, 9475512, 9475513 |
70.20 €
Pack of 50 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
10012223
|
Hirschmann™
9470013 |
Labopette models 9475406, 9475407, 9475503, 9475504, 9475505, 9475506, 9475602, 9475702 |
70.55 €
Pack of 50 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
10052173
|
Hirschmann™
9470012 |
Labopette models 9475408, 9475507, 9475604, 9475704 |
70.55 €
Pack of 50 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
10173913
|
Hirschmann™
9470011 |
Labopette models 9480104, 9480105, 9480106, 9475409, 9475508, 9475509, 9475510, 9480800 |
70.55 €
Pack of 50 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
Description
- Provides the decisive advantage when protecting pipettes and samples against contamination
- Extends the service life of pipettes
- Provides excellent protection for the internal components if replaced regularly
- Can serve as additional protection against penetration of liquids into the interior of the pipette
- For use with a range of Labopette™ models
Specifications
Safety filter | |
Protection against cross-contamination |