missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Safe Imager™ 2.0 Blue-Light Transilluminator

Product Code. 10507584
Change view
Click to view available options
Dimensions (L x W x H):
29.5 x 32.5 x 6.5 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
10507584 29.5 x 32.5 x 6.5 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10507584 Supplier Invitrogen™ Supplier No. G6600EU
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Safe Imager™ 2.0 Blue-Light Transilluminator

The Safe Imager™ Blue-Light Transilluminator is designed for viewing stained gels on the laboratory bench top, or within gel documentation systems. Light from the LED source inside the instrument passes through a blue filter producing light with a narrow emission peak centered at approximately 470 nm, effective for the excitation of SYBR™ DNA-binding dyes such as SYBR™ Safe DNA gel stain, as well as many of our protein gel stains such as SYPRO™ Ruby, SYPRO™ Orange, and Pro-Q™ Diamond stains. Sensitivity obtained using this instrument is comparable to that obtained with a standard UV transilluminator. Unlike UV transilluminators, the Safe Imager™ transilluminator does not produce UV light and does not require UV protective equipment during use.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Transilluminator
For Use With (Application) Nucleic acid gel electrophoresis and blotting, protein gel electrophoresis
For Use With (Equipment) Safe Imager™
Dimensions (L x W x H) 29.5 x 32.5 x 6.5 cm
Electrical Requirements Plug type: EU
Content And Storage • Safe Imager
• Amber filter unit
• Power cord
• Safe Imager viewing glasses

Shipping Condition Room Temperature
Green Features Energy efficient
Lighting Type LED Blue

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.