Learn More
Ansell Edmont™ Safe-Knit™ GP 72-265 Series Grey Polyester/Glass Fiber Heavy Weight Gloves
Description
- Safe-Knit™ GP sleeves contain a higher proportion of Dyneema™ fibers which are as strong as steel fiber but much lighter and more flexible.
- Safe-Knit™ GP Sleeves provide optimal protection from cuts and abrasions, while their light weight and comfortable fit ensure high levels of user acceptance.
Specifications
Specifications
| Back Material | Polyester/Glass Fiber |
| Certifications/Compliance | Standard CATEGORY II EN 388 : 354X |
| Cuff Material | Polyester/Glass Fiber |
| Product Type | Heavyweight Gloves |
| Glove Cuff Style | Attached Sleeve |
| Liner Material | Dyneema™ |
| Lining | Dyneema™ |
| Quantity | 72 Pairs |
| Resistant To | Cut resistant |