missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Safe Lock Micro Test Tube
GREENER_CHOICE

Product Code. 10768371 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Blue
Green
Red
Yellow
Capacity (Metric):
0.5 mL
2 mL
Unit Size:
Pack of 1000
Pack of 500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Capacity (Metric) unitSize
10768371 Green 0.5 mL Pack of 500
10663981 Blue 2 mL Pack of 1000
10058770 Red 2 mL Pack of 1000
10547381 Yellow 2 mL Pack of 1000
10421193 Yellow 0.5 mL Pack of 500
10490993 Blue 0.5 mL Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10768371 Supplier Eppendorf™ Supplier No. 0030121147
  • ONLINE EXCLUSIVE PRICE
    70.20 €
    68.90 € / Pack of 500
    Save 1.30 €
    1% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

Use these polypropylene micro tubes for a variety of applications. Eppendorf™ Safe Lock Micro Test Tubes feature hinged Eppendorf Safe-Lock lids for outstanding protection against unintentional opening during incubation and storage.

  • Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage
  • Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without mold release agents, softeners or biocides that could have a negative effect on bioassays
  • g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Precise lid sealing for minimal evaporation rates during long-term storage
  • Large frosted lid and surface on the side for easy labeling
  • Guaranteed functionality from -86 to +100°C
  • Autoclavable when open (121°C, 20 min.)
  • Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur quality for guaranteed purity
  • Available in Eppendorf Protein and DNA LoBind material for maximum recovery of valuable samples
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 0.5 mL
Color Green
Material Polypropylene
Packaging 500/Pk.
Temperature Range (Metric) –86 to 100°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.