missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ SafeFeet Polypropylene Overshoes with SkidGuard

Product Code. 17790149 Shop All Medicom Products
Change view
Click to view available options
Color:
White
White, Green
White, Pink
Size:
X-Large
2X-Large
Unit Size:
Pack of 300
Pack of 400
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Size Unit Size
17790149 White, Green X-Large Pack of 400
17780149 White, Pink X-Large Pack of 400
17700159 White 2X-Large Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17790149 Supplier Kolmi Hopen™ Supplier No. H06751B
  • ONLINE EXCLUSIVE PRICE
    100.11 €
    79.58 € / Pack of 400
    Save 20.53 €
    20% Off
View all promotions
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Help prevent falls in the workplace with these skid-resistant, polypropylene overshoes.

  • Waterproof and non-slip soles
  • Breathable and lightweight
  • Elastic ankle support
  • Non-woven polypropylene with polyethylene soles
  • Made in France

Specifications

Size X-Large
Material Polypropylene
Color White, Green
Disposable Single-use
Thickness 60 μm
For Use With (Application) Medical, Industrial
Certifications/Compliance ISO 13485, (EU) 2017/745
Weight (Metric) 34 g/m2
Quantity 400 Pk., 400 Packungen, 400 paq.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.