Learn More
Brady™ SafeKey Padlocks - aluminum with 76 mm Steel Shackle, Keyed Different
The ultimate locks for indoor and outdoor heavy duty safety applications.
35.65 € - 218.00 €
Specifications
Product Type | Keyed Padlocks |
---|---|
Key Type | Keyed Different |
Material | Anodized Aluminum |
Diameter (English) Shackle | 0.25 in. |
Diameter (Metric) Shackle | 6.4 mm |
Product Code | Brand | Quantity | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Color | Price | Quantity & Availability | ||||
17160291
|
Brady™
150241 |
1 Ea. | Blue |
35.65 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
17170321
|
Brady™
150283 |
1 Ea. | Silver |
35.65 €
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
17180321
|
Brady™
150284 |
1 Ea. | Brown |
35.65 €
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
17190321
|
Brady™
150285 |
1 Ea. | Yellow |
35.65 €
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
17170341
|
Brady™
150306 |
1 Ea. | Orange |
35.65 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
17150361
|
Brady™
150330 |
1 Ea. | Purple |
35.65 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
17160361
|
Brady™
150331 |
1 Ea. | Black |
35.65 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
17170361
|
Brady™
150332 |
1 Ea. | Red |
35.65 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
17160381
|
Brady™
150360 |
1 Ea. | Green |
35.65 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
17160271
|
Brady™
150216 |
6 Pk. | Black |
210.00 €
Pack of 6 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
17120291
|
Brady™
150235 |
6 Pk. | Brown |
210.00 €
Pack of 6 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
17150301
|
Brady™
150252 |
6 Pk. | Green |
210.00 €
Pack of 6 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
17190331
|
Brady™
150297 |
6 Pk. | Silver |
210.00 €
Pack of 6 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
17110361
|
Brady™
150322 |
6 Pk. | Blue |
210.00 €
Pack of 6 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
17120361
|
Brady™
150323 |
6 Pk. | Red |
218.00 €
Pack of 6 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
17160371
|
Brady™
150344 |
6 Pk. | Orange |
210.00 €
Pack of 6 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
17170371
|
Brady™
150345 |
6 Pk. | Yellow |
210.00 €
Pack of 6 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
17150381
|
Brady™
150358 |
6 Pk. | Purple |
210.00 €
Pack of 6 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
Description
- Manufactured with solid aluminum body and steel shackle, Brady™'s padlocks are lightweight and rugged
- Compliments the Brady™ SafeKey Padlock range and have the same features
- Locks are chemical and corrosion resistant
- The unique lock mechanism is insulated and has 6 precision steel tumbler blades for a high number of key differences and tamper resistance
- The colour-matched key and lock body allows a fast identification in the application
- 1 key is supplied with each lock
- Custom keying, engraving, charting, Master Key and Grand Master Key are available
Spécification
Keyed Padlocks | |
Anodized Aluminum | |
6.4 mm | |
76 mm | |
6 | |
1.3 in. | |
33 mm |
Keyed Different | |
0.25 in. | |
3 in. | |
Steel | |
1.5 in. | |
38 mm |