Learn More
DuPont™ Tyvek 500 Xpert Hooded Coverall
Product Code. 12911355
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
Unit Size:
Each
Pack of 100
Pack of 25
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12911355 Large Pack of 25
11743103 Small Pack of 100
12971345 Small Pack of 25
13015683 Small Each
13412429 Medium Pack of 25
12991345 Medium Pack of 25
11703103 Medium Pack of 100
13005683 Medium Each
13095673 Large Each
11713103 Large Pack of 100
11723103 X-Large Pack of 100
12931355 X-Large Pack of 25
13025683 X-Large Each
12951355 2X-Large Pack of 25
13075673 2X-Large Each
11733103 2X-Large Pack of 100
12961355 3X-Large Pack of 25
11753103 3X-Large Pack of 100
13085673 3X-Large Each
17402893 4X-Large Pack of 100
17412893 5X-Large Pack of 100
17422893 6X-Large Pack of 100
17432893 7X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Product Code. 12911355 Supplier DuPont™ Supplier No. D14663977
  • ONLINE EXCLUSIVE PRICE
    289.00 €
    213.75 € / Pack of 25
    Save 75.25 €
    26% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Composed of flash spun high density polyethylene. DuPont™ Tyvek™ Classic Xpert Hooded Coverall is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
Composed of flash spun high density polyethylene. Dupont™ Tyvek™ Classic Xpert Hooded Coverall is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
  • Robust yet lightweight (<180g per garment)
  • Elasticated face, wrists and ankles as well as glued-in waist elastic
  • Ample crotch area for freedom of movement
  • Large, easy-to-grasp zipper puller
  • 3-piece hood for optimal fit to head and face when turning
  • White, sizes S to XXXL
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size Large
Material Flashspun HDPE
Color White
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Self-locking Zipper with Flap
Seam Type Stitched
Certifications/Compliance Category III, Type 5-B and 6-B, EN 14126, EN 1073-2, EN 1149-5
Size (Metric) Chest 100 to 108 cm
For Use With (Application) Pharmaceutical handling, chemical processing, the oil and gas industry, general maintenance and operations, automatic spray painting and many others.
Quantity 100/Pk.
Style Classic Xpert
Show More Show Less
The user should make the risk assessment to determine the protection required.