Learn More
DuPont™ Tyvek 500 Xpert Hooded Coverall
Product Code. 17432893 Shop All DuPont Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
Unit Size:
Each
Pack of 100
Pack of 25
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17432893 7X-Large Pack of 100
13015683 Small Each
11743103 Small Pack of 100
12971345 Small Pack of 25
11703103 Medium Pack of 100
13412429 Medium Pack of 25
13005683 Medium Each
12991345 Medium Pack of 25
13095673 Large Each
12911355 Large Pack of 25
11713103 Large Pack of 100
11723103 X-Large Pack of 100
12931355 X-Large Pack of 25
13025683 X-Large Each
12951355 2X-Large Pack of 25
13075673 2X-Large Each
11733103 2X-Large Pack of 100
12961355 3X-Large Pack of 25
11753103 3X-Large Pack of 100
13085673 3X-Large Each
17402893 4X-Large Pack of 100
17412893 5X-Large Pack of 100
17422893 6X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Product Code. 17432893 Supplier DuPont™ Supplier No. D15553616
  • ONLINE EXCLUSIVE PRICE
    1342.89 €
    990.16 € / Pack of 100
    Save 352.73 €
    26% Off
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Composed of flash spun high density polyethylene. DuPont™ Tyvek™ Classic Xpert Hooded Coverall is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
Composed of flash spun high density polyethylene. Dupont™ Tyvek™ Classic Xpert Hooded Coverall is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
  • Robust yet lightweight
  • 3-piece hood for optimal fit to head and face when turning
  • Elasticated face, wrists and ankles as well as glued-in waist elastic
  • Ample crotch area for freedom of movement
  • Large, easy-to-grasp zipper puller
  • Very low inward leakage thanks to optimized design
TRUSTED_SUSTAINABILITY
Specifications
Size 7X-Large
Color White
Closure Type Zipper
Seam Type Stitched
Certifications/Compliance (Europe) (EU) 2016/425, EN 14126, EN 1149-5
The user should make the risk assessment to determine the protection required.