Learn More
WLD Tec™ Safety Adapter
Product Code. 12812184
Change view
Click to view available options
For Use With (Equipment):
Gasprofi 1 SCS Series
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
12812184 Gasprofi 1 SCS Series Each
1 options
Product Code. 12812184 Supplier WLD Tec™ Supplier No. 6.000.750
  • ONLINE EXCLUSIVE PRICE
    109.20 €
    85.80 € / Pack of 1
    Save 23.40 €
    21% Off
Estimated Shipment: 06-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Adapter
For Use With (Equipment) Gasprofi 1 SCS Series
Includes Integrated Pressure Regulator 50 mbar For Gas Cartridge
No. per Pack 1 Ea.