Learn More
Justrite™ Safety Cabinet Slimline
Product Code. 11975426
Change view
Click to view available options
Door Style:
Self-close
Unit Size:
Each
Product Code. Door Style Unit Size
11975426 Self-close Each
1 options
Product Code. 11975426 Supplier Justrite™ Supplier No. 8922221
  • ONLINE EXCLUSIVE PRICE
    2619.75 €
    2110.63 € / Pack of 1
    Save 509.12 €
    19% Off
 Restricted Item
Estimated Shipment: 25-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
1 door, self close, not for phenol, nitric /sulphuric acid, steel, double wall, epoxy powder finish, dual vents, earth wire connections, leakproof sills, levelling feet blue 1,651mm x 591mm x 457mm (w x h x d) 83L
Specifications
Door Style Self-close