missing translation for 'onlineSavingsMsg'
Learn More

SCAT Safety Canisters

Product Code. 10589765 Shop All SCAT Europe Products
Change view
Click to view available options
Capacity (Metric):
2.5 L
5 L
10 L
12 L
20 L
30 L
60 L
Closure Size:
GL45
S50
S55
S60/61
S60/61 and GL45
S70/71
S90
S95
Includes:
2 openings
Floater
Mechanical Level Control
UN-X approval
UN-X approval; with inner fluorination
UN-Y approval
UN-Y approval, with inner fluorination
Unit Size:
Each
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size Includes Unit Size
10589765 5 L GL45 UN-Y approval Each
10292940 2.5 L GL45 UN-Y approval Each
10214825 10 L GL45 UN-Y approval Each
10304460 5 L S50 UN-Y approval Each
15767762 5 L S55 UN-X approval; with inner fluorination Each
17059693 5 L S55 UN-X approval Each
10521011 10 L S55 UN-Y approval Each
10190040 5 L S55 UN-Y approval Each
17069693 5 L S60/61 UN-Y approval Each
17967250 12 L S60/61 Mechanical Level Control Each
17997260 20 L S60/61 - Each
17181511 10 L S55 UN-Y approval, with inner fluorination Each
10588510 12 L S60/61 UN-Y approval Each
10531011 20 L S60/61 UN-Y approval Each
10334840 20 L S60/61 Floater Each
10620441 30 L S60/61 UN-Y approval Each
15531856 30 L S60/61 and GL45 2 openings Each
10765392 60 L S70/71 UN-Y approval Each
10512775 10 L S90 UN-Y approval Each
10704072 5 L S95 UN-Y approval Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
Product Code. 10589765 Supplier SCAT Supplier No. 107951
Big Savings for Better Separation
Use promo code 27992
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Our SCAT Safety Canisters are made of high-quality PE-HD material with high chemical resistance for use in the laboratory for safe disposal of chromatographic solvents or other liquid laboratory waste for the highest level of work safety in the laboratory.

  • Made of robust HDPE, which is very resistant to almost all solvents commonly used in the laboratory
  • Smallest canisters have a volume of only 5 L and are ideally suited for use on tables or smaller storage areas
  • Larger canisters are available with volumes of 10, 12, 20, 30 up to 60 L, which can be deposited under tables or in safety cabinets
  • Thread types S55, S60/61, S65, S70/71, S90 and S95 are commonly used in the laboratory, they are all compatible with the SCAT SafetyWasteCaps range for the safe disposal of volatile hazardous substances
  • Special versions available with floats for level indication or with internal fluorination for double protection of the contents, both for permeation from outside to inside and for diffusion from inside to outside

The wide range of non-electrically conductive canisters includes a variety of designs for individual use in the laboratory for the disposal of solvents in combination with the SCAT SafetyWasteCaps to increase your laboratory safety.

Specifications

Autoclavable Not Autoclavable
Capacity (English) 1.3 gal.
Capacity (Metric) 5 L
Certifications/Compliance UN-Y Approval
Closure Size GL45
Color White (Translucent)
Description Canister 5 L, GL45, PE-HD, UN-Y approval, dimensions (W x H x D): 150 x 250 x 195 mm
Dimensions (L x W x H) 195 x 150 x 250 mm
For Use With (Application) For All Purposes and Suitable for The S.C.A.T™ SafetySystem
Height (Metric) 365 mm
Includes UN-Y approval
Material HDPE
Mouth Style GL45 Thread
Product Type Canister
Sterility Non-sterile
Width (Metric) 150 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.