missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAT HPLC Safety Caps
These safety caps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapors. Complete with integrated air valve for pressure equalization.
131.00 € - 328.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
For Use With (Application) | HPLC Systems |
Material | PTFE |
Product Code | Brand | For Use With (Equipment) | Includes | Thread Size (Metric) | Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Includes | Thread Size (Metric) | Type | Price | Quantity & Availability | ||
10851421
|
SCAT
107030 |
Lab Bottles | 1 PFA Fitting with 3.2 mm OD, 1 Air Valve | GLS80 | Safety Cap I |
131.00 €
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
10348149
|
SCAT
107607 |
Ground Neck Bottles 29/32 mm | 2 PFA Fittings with 3.2 mm OD, 1 Air Valve | NS29/32 | Safety Cap II |
157.00 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||
10541728
|
SCAT
107031 |
Lab Bottles | 2 PFA Fittings with 3.2 mm OD, 1 Air Valve | GLS80 | Safety Cap II |
144.00 €
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
16554170
|
SCAT
107742 |
Lab Bottles | 3 PFA Fittings with 3.2 mm OD, 1 Air Valve | GLS40 | Safety Cap III |
144.00 €
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
15168299
|
SCAT
107512 |
Lab Bottles | 3 PFA Fittings with 1.6/2.3/3.2 mm OD, 2 PFA Blind Plugs, 1 Air Valve | GL38 | Safety Cap III |
132.00 €
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
10839170
|
SCAT
107032 |
Lab Bottles | 3 PFA Fittings with 3.2 mm OD, 1 Air Valve | GLS80 | Safety Cap III |
157.00 €
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
10222440
|
SCAT
107035 |
Lab Bottles | 4 PFA Fittings with 3.2 mm OD, 1 Air Valve | B83 | Safety Cap IV |
328.00 €
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||
Description
- High quality materials ensure maximum safety for lab and staff
- 360° rotatable and up to 200°C autoclavable screw cap for maximum flexibility
- Stable construction for highest safety and easy handling
- Chemically resistant against aggressive organic solvents
- Attractive design and clear arrangement of the connections
- Improved design for easy connection of capillaries in various colors
- Free air valve with change indicator included with every SafetyCap
Specifications
Autoclavable | |
HPLC Systems | |
PTFE |