Learn More
SCAT HPLC Safety Caps, V2.0, with Shut-Off
SafetyCap is designed to safety extract solvents when working with any HPLC/UHPLC system. The SCAT Safety Caps provide maximum safety for your HPLC solvent supply. Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours.
Brand: SCAT 307919
Additional Details : Weight : 0.10900kg
Description
- Luer Lock Adapter
- Improved fitting design for easier connection
- Constructed of medical grade PTFE, PFA fittings, and a PPS screw cap
- Includes space for individually labeling
- Autocalvable up to 200°C
- Stable construction for improved handling
Specifications
2 fittings, 1 air valve, 2 shut-offs | |
HPLC Systems | |
PFA, PTFE, PPS | |
GL 45 |
3.2 mm O.D. fitting | |
Safety Cap | |
Lab Bottles |