missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Safety Clip Conical Joint Clips

Product Code. 11963361
Change view
Click to view available options
For Use With (Equipment):
For ground joint NS 24/29
For ground joint NS 45/40
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11963361 For ground joint NS 45/40 Pack of 10
11933361 For ground joint NS 24/29 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11963361 Supplier Lenz Laborglasintrumente™ Supplier No. 01103845
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

POM safety clip for conical ground joints

  • Height adjustable by variable gear rods, also for use with sleeves with grip collar
  • Outstanding chemical resistance
  • Operating temperature up to approximately 150°C

Specifications

Quantity 10
Material POM
For Use With (Equipment) For ground joint NS 45/40
Color Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.