missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Safety Cover for Heat-On Multi-Well Holder

Product Code. 13400039 Shop All Heidolph Instruments Products
Change view
Click to view available options
For Use With (Equipment):
Heat-On Multi-Well Holder
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
13400039 Heat-On Multi-Well Holder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13400039 Supplier Heidolph™ Supplier No. 5058008000
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This cover provides an extra level of protection against burns when utilizing the Heat-On Multi-Well holder.

  • Ensures more protection against burns when using Heat-On Block
  • Despite reduced danger when using Heat-On Blocks as compared to conventional heating bath, risk of burns is not zero- this safety cover reduces that
  • Cover is made of chemical-resistant, easy-to-clean, PTFE
  • Only suitable for the Multi-Well holder that encloses Heat-On inserts
TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Equipment) Heat-On Multi-Well Holder
For Use With (Application) Heat Protection
Material PTFE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.