missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Safety Pipetting Ball

Product Code. 10496263 Shop All Bürkle Products
Click to view available options
Color:
Blue
Red
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10496263

Brand: Buerkle™ 75540002

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Peleus ball, simple pipetting aid. Evacuation takes place via an automatic valve. By pressing the valves, the liquid is drawn up, dispensed or purged. The removable automatic valve also enables effective cleaning of the inside

  • For all pipettes up to 100 ml
  • Available in various colours
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Type Ball
For Use With (Equipment) up to 100 mL Pipettes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.