Learn More
Pacific Handy Cutter Safety Point Blades Dispenser Unit
Product Code. 17709047
Change view
Click to view available options
Product Type:
Blade Dispenser
Unit Size:
Pack of 6
Product Code. Product Type Unit Size
17709047 Blade Dispenser Pack of 6
1 options
Product Code. 17709047 Supplier Pacific Handy Cutter Supplier No. SPD017
  • ONLINE EXCLUSIVE PRICE
    283.00 €
    264.22 € / Pack of 6
    Save 18.78 €
    6% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
The SPD-017 Blade Dispenser safely dispenses one SP-017 Safety Point Blade at a time from a durable plastic case by easily pulling the yellow tab.
  • This convenient gravity fed dispenser helps control blade inventory and insures that employees have the proper tools where and when needed to remain productive.
  • The dispenser contains 100 new blades and fits most PHC safety cutters including the S4, S4S, S5, S7, S8, EZ1, EZ2+, EZ4, EZ7, GSC3, RZ3 and EZ3.
  • Plus, by pairing a PHC Blade Dispenser (SPD-017, QBD-324, or QBD-379) to the front of the PHC Blade Bank you create a blade change station that will increase the safety and efficiency of your staff.
  • By using both items together your employees will be encouraged to properly dispose of loose blades and easily access new replacement blades near their work stations and throughout your facility, ultimately increasing overall productivity. 
Specifications
Product Type Blade Dispenser
For Use With (Application) Easily Access New Replacement Blade
Disposable Single-use
Sterility Non-sterile