missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Cabinet for Acids and Alkalis SL-CLASSIC

Product Code. 15231035 Shop All asecos Products
Change view
Click to view available options
Dimensions (D x W x H) Interior:
570 x 485 x 914 mm
570 x 485 x 1869 mm
No. of Shelves:
4
6
8
None
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (D x W x H) Interior No. of Shelves unitSize
15231035 570 x 485 x 1869 mm None Each
12643515 570 x 485 x 914 mm 6 Each
15221035 570 x 485 x 1869 mm 8 Each
11959636 570 x 485 x 914 mm 4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy

For indoor use only

Product Code. 15231035 Supplier asecos™ Supplier No. 3060600130608

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For indoor use only

Safe and approved storage of aggressive, non flammable hazardous materials in working areas.

Safe and approved storage of aggressive, non flammable hazardous materials in working areas

Function / construction

  • Robust construction and longevity: cabinet body made of powder-coated sheet steel, safety elements assembled outside the storage compartment for increased protection against corrosion
  • Two in one: two hermetically separated storage compartments allow the isolated storage of acids and bases in one cabinet
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • ventilation: integrated metal-free air ducts ready for connection to a technical exhaust system with only one extraction air inlet

Certificates

  • type tested in accordance with EN 14727
  • EN 16121/16122 conformity
  • GS certified

Warranty

Five years

Specifications

Certifications/Compliance DIN EN 16121/16122 conformity, GS certified
Configuration 12 x pull-out shelf (panel melamine resin-coated/PP)
Depth (Metric) Exterior 603 mm
Depth (Metric) Interior 570 mm
Dimensions (D x W x H) Interior 570 x 485 x 1869 mm
Dimensions (D x W x H) 603 x 1197 x 1965 mm
Door Style Wing
For Use With (Application) Safe and approved storage of aggressive, non fla mmable hazardous materials in working areas
Height (Metric) Exterior 1965 mm
Height (Metric) Interior 1865 mm
Model SL.196.120.MV
No. of Doors 2
No. of Shelves None
Shelf Type Pull-out
Surface Coating Melamine resin laminated
Weight (Metric) 291.3 kg
Width (Metric) Exterior 1197 mm
Width (Metric) Interior 485 mm
Color Light Gray
Depth (English) Exterior 23.74 in.
Height (English) Exterior 77.36 in.
Material Chipboard, Polypropylene
Product Type Acids and Alkalis Cabinet
Width (English) Exterior 47.13 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.