Learn More
SCAT Safety Waste Cap 'LISA', V3.0 Blind Plug
Safety Waste Cap 'LISA', V3.0 Blind Plug is an accessory suitable for safely closing unused threaded holes in devices for the safe disposal of liquid HPLC/UHPLC waste.
Brand: SCAT 450110
46.70 EUR valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.31100kg
Description
- Accessory for Safety Waste Cap 'LISA,' V3.0
- For extending and attaching various components such as a funnel
- Made of PTFE for optimal chemical resistance
Specifications
Satellite Blind Plug | |
PTFE | |
M30x35 |
Electrically Conductive | |
HPLC / UHPLC Systems |