Learn More
SCAT Safety Waste Cap 'LISA', V3.0 Blind Plug
Shop All SCAT Europe Products
missing translation for 'orderingAttributeHoverText'
Type:
Electrically Conductive
Not Electrically Conductive
Thread Size:
GL14
M30x35
NPT 1/8 in.
Includes:
Blind plug
Satellite Blind Plug
missing translation for 'unitSize'
Each
Description
- Accessory for Safety Waste Cap 'LISA,' V3.0
- For extending and attaching various components such as a funnel
- Made of PTFE for optimal chemical resistance
Specifications
Specifications
| Type | Electrically Conductive |
| Includes | Blind plug |
| Thread Size | GL14 |
| For Use With | HPLC / UHPLC Systems |
| Material | PTFE |