Learn More
SymLine™ Safety Waste Caps
These Safety Waste Caps safely connect the pipework to the waste container as an integrated exhaust filter blocks solvent vapors for health and environmental protection.
87.90 € - 730.00 €
Product Code | Brand | Thread Size | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Includes | Price | Quantity & Availability | ||||
17145501
|
SymLine™
306497 |
GL45 | 2 x PFA-fittings for Capillaries with (OD) 1.6/2.3/3.2 mm, Tube Connector for (ID) 5 to 11.5 mm, Collector for HPLC Waste |
97.65 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
17135501
|
SymLine™
306491 |
GL45 | 3 x PFA-fittings for Capillaries with (OD) 1.6/2.3/3.2 mm, Collector for HPLC Waste |
87.90 €
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
16876011
|
SymLine™
306494 |
GL45 | 5 x PFA-fittings for Capillaries with (OD) 1.6/2.3/3.2 mm, Collector for HPLC Waste |
114.00 €
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||||
15960386
|
SymLine™
106478 |
S55 | Electronic Level Control, GL25 (m), Immersed Filling Lance, Connector for Exhaust Filter |
398.00 €
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
15724922
|
SymLine™
106522 |
S55 | 1 x GL25 (m), Immersed Filling Lance, Connector for Exhaust Filter |
182.00 €
Each |
Estimated Shipment: 25-07-2024
Log in to see stock. |
||||
17808743
|
SymLine™
106719 |
S55 | Electronic Level Control with Shut-off GL25 (m), Connector for Exhaust Filter |
575.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
17079693
|
SymLine™
306581 |
S60/61 | Adjustable PTFE Level Control, Cable Connection (2 m), Connector for Exhaust Filter |
600.00 €
Each |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||||
16866011
|
SymLine™
306482 |
S60/61 | 2 x PFA-fittings for Capillaries with (OD) 1.6/2.3/3.2 mm, Tube Connector for (ID) 5 to 11.5 mm |
259.00 €
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17813352
|
SymLine™
106718 |
S60/61 | Electronic Level Control with Shut-off GL25 (m), Connector for Exhaust Filter |
575.00 €
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17377409
|
SymLine™
106484 |
S90 | Electronic Level Control, GL25 (m), Connector for Exhaust Filter |
525.00 €
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17838743
|
SymLine™
106720 |
S90 | Electronic Level Control with Shut-off GL25 (m), Connector for Exhaust Filter |
730.00 €
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
18646478
|
SymLine™
307971 |
S60/61 | 1x NPT 3/8 in. (f), 1x connection for exhaust filter (45° beveled) |
219.55 €
1 sheet |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Available with different thread sizes for attachment to pipe system
- Different numbers of hose connections for disposal of liquid waste
- Available with electronic level sensors that indicate the degree of filling of the disposal container
- By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system