missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SymLine Safety Waste Collector
Shop All SCAT Europe Products
Click to view available options
Includes:
2 x PFA-fittings
2 x PFA-fittings, 1 tube connector
3 x PFA-fittings
4 x PFA-fittings
4 x PFA-fittings, 1 tube connector
5 x PFA-fittings
7 x PFA-fittings
Unit Size:
1 set
Description
SymLine Safety Waste Collectors route the waste tubes from your HPLC system directly into the disposal system. A choice of several connections offers you complete freedom. Fittings for different capillary diameters are included in the scope of delivery.
- Made of electrically conductive PTFE-EL material to prevent static discharge
- Available with different numbers of tube connections
- For use on a SymLine pipe system for liquid waste disposal
Specifications
Specifications
| For Use With (Equipment) | Capillaries with (OD) 1.6/2.3/3.2 mm |
| Includes | 2 x PFA-fittings |
| Material | PTFE-EL |
| Thread Size | GL45 |
| Type | Safety Waste Collector |
| Color | Black |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction