missing translation for 'onlineSavingsMsg'
Learn More

SymLine Safety Waste Collector

Product Code. p-7250698 Shop All SCAT Europe Products
Click to view available options
Includes:
2 x PFA-fittings
2 x PFA-fittings, 1 tube connector
3 x PFA-fittings
4 x PFA-fittings
4 x PFA-fittings, 1 tube connector
5 x PFA-fittings
7 x PFA-fittings
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17895704

Brand: SymLine 306489EL

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

SymLine is the modular disposal system for liquid waste. It connects several laboratory workstations, HPLC systems and fume cupboards to a central disposal or collection point, such as a safety cabinet. Made of electrostatic conductive PTFE connects capillaries and tubes of the HPLC system with the disposal system.

SymLine Safety Waste Collectors route the waste tubes from your HPLC system directly into the disposal system. A choice of several connections offers you complete freedom. Fittings for different capillary diameters are included in the scope of delivery.

  • Made of electrically conductive PTFE-EL material to prevent static discharge
  • Available with different numbers of tube connections
  • For use on a SymLine pipe system for liquid waste disposal

Specifications

For Use With (Equipment) Capillaries with (OD) 1.6/2.3/3.2 mm
Includes 2 x PFA-fittings
Material PTFE-EL
Thread Size GL45
Type Safety Waste Collector
Color Black
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.