missing translation for 'onlineSavingsMsg'
Learn More

Sartorius SafetySpace™ Filter Pipet Tips
BIOPHARMAGREENER_CHOICE

Product Code. 13197873 Shop All Sartorius Products
Change view
Click to view available options
Color Code:
Blue
Gray
Orange
Purple
Yellow
Length (Metric):
3.2 cm
4.6 cm
5.1 cm
5.25 cm
7.8 cm
9cm
Volume (Metric):
0.1 to 10 μL
2 to 120 μL
5 to 1000 μL
5 to 200 μL
5 to 300 μL
50 to 1200 μL
Unit Size:
Pack of 960
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Code Length (Metric) Volume (Metric) unitSize
13197873 Yellow 5.1 cm 2 to 120 μL Pack of 960
10587600 Gray 3.2 cm 0.1 to 10 μL Pack of 960
10014554 Yellow 5.25 cm 5 to 200 μL Pack of 960
13107634 Orange 5.25 cm 5 to 300 μL Pack of 960
11894782 Purple 9cm 50 to 1200 μL Pack of 960
10708262 Gray 4.6 cm 0.1 to 10 μL Pack of 960
11552934 Blue 7.8 cm 5 to 1000 μL Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 13197873 Supplier Sartorius Supplier No. 790101F
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Provides more space between the sample and the filter than conventional filter tips which virtually eliminates the risk of an expensive or a contagious sample permeating the filter. Sartorius SafeySpace™ Filter Pipet Tips allow pipetting any type of liquid or using any pipetting technique without the risk of the sample absorbing into the filter.

SafetySpace™ Filter Tips leave more space between the sample and the filter than conventional filter tips. This allows pipetting any type of liquid or using any pipetting technique without the risk of the precious sample absorbing into the filter. In effect, this secures 100% sample recovery and thus greater accuracy.

  • Polypropylene
  • With regular air gap and filter barrier
  • Single tray packaging
  • Purity level: RNase, DNase, Pre-endotoxin-free sterilized

The safety space feature is particularly useful in:

  • pipetting foaming liquids such as buffers and proteins
  • pipetting solvents
  • multiple dispensing functions of electronic pipettes
  • reverse pipetting
  • any other application requiring extra space in the tip

The tips are made of virgin polypropylene and the filter of polyethylene without 'self-sealing' additives to avoid any interference with the sample and the results. The filter barrier helps protect against cross-contamination of the sample and/or the pipette and reduces the need for pipette maintenance. The filter tips are available in multiple volumes from 10 μl to 1200 μl and are packed in colour coded single tray racks. They are DNase, RNase and endotoxin free certified and pre-sterilized.

The filter barrier and the certified purity makes the filter tips ideal for:

  • molecular biology
  • microbiology
  • cell culture applications
  • radioactive work
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 2 to 120 μL
Filtered Filtered
Sterility Sterile
Color Colorless
Autoclavable Not Autoclavable
Length (English) 2 in.
Length (Metric) 5.1 cm
Certifications/Compliance CE, ISO, IVD
Clean Claims DNase, RNase, and Endotoxin Free
Sterilization Method Pre-sterilized
Format Rack
Product Type Pipette Tips
Color Code Yellow
Disposable Single-use
For Use With (Equipment) Sartorius, Universal Fit
Graduated No
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.