Learn More
HALYARD™ SAFEVIEW™ Protective Eyewear, Frames Only
Product Code. 30274745 Shop All Halyard Products
Change view
Click to view available options
Quantity:
100
Unit Size:
Pack of 100
Product Code. Quantity Unit Size
30274745 100 Pack of 100
1 options
Product Code. 30274745 Supplier HALYARD™ Supplier No. SV100F
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SAFEVIEW™ Eyewear protects eyes in splashing or spraying environments. Lightweight and latex-free, these glasses comfortably fit over most prescription eyewear and come in various colorful frames.
  • HALYARD™ SAFEVIEW™ Eyewear provides reliable eye protection for healthcare professionals and other users in environments where splashing or spraying may occur.
  • For use in situations which may involve splashing or spraying
  • Certified as PPE Category II, for moderate risks
  • Lightweight design to reduce strain and increase comfort in use
  • Colorful frames appeal to a variety of preferences
  • Designed to be easily worn over most prescription glasses
  • Constructed from natural rubber latex-free and DEHP-free materials
  • Lenses film: clear polyester; Frames: polypropylene
Specifications
Certifications/Compliance PPE Regulation 2016/425 (PPE category II)
Color Clear
Product Type Replacement Frame
Frame Color Assorted colours
No. per Case 100
Quantity 100
No. per Pack 10
Size One-size