Learn More
HALYARD™ SAFEVIEW™ Protective Eyewear, Replacement Lenses
Product Code. 30274676 Shop All Halyard Products
Change view
Click to view available options
Quantity:
250
Unit Size:
Pack of 250
Product Code. Quantity Unit Size
30274676 250 Pack of 250
1 options
Product Code. 30274676 Supplier HALYARD™ Supplier No. SV250L
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SAFEVIEW™ Eyewear protects eyes in splashing or spraying environments. Lightweight and latex-free, these glasses comfortably fit over most prescription eyewear and come in various colorful frames.
  • HALYARD™ SAFEVIEW™ Eyewear provides reliable eye protection for healthcare professionals and other users in environments where splashing or spraying may occur.
  • For use in situations which may involve splashing or spraying
  • Certified as PPE Category II, for moderate risks
  • Lightweight design to reduce strain and increase comfort in use
  • Colorful frames appeal to a variety of preferences
  • Designed to be easily worn over most prescription glasses
  • Constructed from natural rubber latex-free and DEHP-free materials
  • Lenses film: clear polyester; Frames: polypropylene
Specifications
Certifications/Compliance PPE Regulation 2016/425 (PPE category II)
Color Clear
Product Type Eyeshield Lens
No. per Case 250
Quantity 250
No. per Pack 25
Size One-size
Disposable Single-use
Autoclavable Not Autoclavable