Learn More
MEDICOM Safewear Low Waterproof Overboots with Sole
Product Code. 31781294 Shop All Medicom Products
Change view
Click to view available options
Size:
Universal
Unit Size:
Pack of 400
Product Code. Size Unit Size
31781294 Universal Pack of 400
1 options
Product Code. 31781294 Supplier MEDICOM Supplier No. 10.202
  • ONLINE EXCLUSIVE PRICE
    222.49 €
    197.92 € / Pack of 400
    Save 24.57 €
    11% Off
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Low waterproof overshoes provide total protection against moisture and grip, even on wet floors.
  • Made of embossed PE, the overboot is 100% waterproof: tested according to the EN ISO 811 water penetration resistance method under pressure
  • The sole provides good grip on the ground, even on wet surfaces
  • Excellent abrasion resistance even during prolonged use: tested according to the EN ISO 12947-2 method
  • Reinforced model in 200 μm embossed PE
  • Its foot-shaped (anatomical) design ensures easy fitting for the user, even in uncomfortable positions
  • The overboot fully covers the user's footwear, fitting sizes from 34 to 48
  • The elastic band ensures excellent ankle support
Specifications
Color White
Material Polyethylene
Size Universal
Gender Unisex
Waterproof Yes
Product Type Overboot