Description
- Attached with resistant expandable PE links
- One by one distribution system to avoid contamination of the entire batch when the apron is removed from its dispenser bag
- Possibility of color coding
- It adapts to different body shapes
- Food contact
Specifications
Specifications
| Type | Apron |
| Size | Universal |
| Material | Polyethylene |
| Color | Blue |
| Disposable | Single-use |
| Closure Type | Adjustable Ties |
| Sterility | Non-sterile |
| Dimensions (L x W) | 120 x 70 cm |