Learn More
MEDICOM Safewear PP Gown 40 g/m2, Zip
Product Code. 31781374 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Unit Size:
Pack of 50
Product Code. Size Unit Size
31781374 Small Pack of 50
1 options
Product Code. 31781374 Supplier MEDICOM Supplier No. 40.635/02
  • ONLINE EXCLUSIVE PRICE
    59.51 €
    49.72 € / Pack of 50
    Save 9.79 €
    16% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The polypropylene hygiene gown is comfortable and lightweight, with elasticated cuffs and zip closure to limit minor soiling risks.
  • Single-use hygiene coats protect the work environment from particles coming from the operator's body (scales, hair, sweat, etc/)
  • They are suitable for HACCP processes
  • They allow the operator to protect themselves from the risks of soiling related to the activity they carry out
Specifications
Size Small
Product Type Isolation Gown
Material Polypropylene
Color Blue
Closure Type Zipper
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic