missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Safranin Stain

Product Code. 12976297
Click to view available options
Quantity:
500mL
1 L
2 L
Unit Size:
2 L
This item is not returnable. View return policy

Product Code. 12976297

Brand: Pro-Lab Diagnostics™ PL.7014

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use for Gram staining of bacteria. Pro-Lab Diagnostics™ Safranin Stain is designed to be ready to use.

  • Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions in a selection of convenient and economical sizes.
  • Manufactured in accordance with all current regulatory requirements
  • CE labelled
  • Strict quality control and filtering techniques prior to release
  • Also available are a range of convenient staining kits for Gram, TB and Cryptosporidium staining

Specifications

Type Safranin
Quantity 2 L
Certifications/Compliance CE marked (98/78/EC)
Format Liquid
For Use With (Application) Gram’s Staining Method for the Initial Differentiation of Gram Positive and Gram Negative Bacteria
Color Dark Red
Composition Ethanol 10 - <25% , Methanol 1 - <2.5% , Glycerol 1 - <2.5%
Storage Requirements Keep at temperature not exceeding 20°C
Stain Type Gram
compliance-icons compliance-icons
Product Identifier
  • Safranin
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501-Dispose of contents/container to .
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash contaminated skin thoroughly after handling.
  • P370+P378-In case of fire: Use foam, carbon dioxide, dry powder or water fog to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.