missing translation for 'onlineSavingsMsg'
Learn More

Safranin T (>75% Purity), TRC

Product Code. 30421274
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421274 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421274 Supplier Toronto Research Chemicals Supplier No. TRCS8190535G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Basic Red 2
CAS 477-73-6
Molecular Formula C20 H19 N4 . Cl
Quantity 5 g
Synonym 3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chloride (1:1), C.I. Basic Red 2, 3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chloride, Safranine O, 2,8-Dimethylphenosafranine, 3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chloride, Basic Pink, Basic Red 2, Brilliant Safranine BR, Brilliant Safranine G, Brilliant Safranine GR, C.I. 50240, Calcozine Red Y, Duasyn Basic Red TH, Gossypimine, Hidaco Safranine, Leather Red HT, Lowacryl Red 2, Lowacryl Red 2 Conc, Mitsui Safranine T, Nippon Kagaku Safranine GK, Nippon Kagaku Safranine T, Safranin, Safranin O, Safranin T, Safranin orange, Safranine, Safranine A, Safranine B, Safranine G, Safranine GF, Safranine J, Safranine OK, Safranine OK 70:100, Safranine Superfine G, Safranine T, Safranine TH, Safranine TN, Safranine TS, Safranine Y, Safranine YN, Safranine Zh, Tolusafranine
InChI Formula InChI=1S/C20H18N4.ClH/c1-12-8-17-19(10-15(12)21)24(14-6-4-3-5-7-14)20-11-16(22)13(2)9-18(20)23-17;/h3-11H,1-2H3,(H3,21,22);1H
SMILES [Cl-].Cc1cc2nc3cc(C)c(N)cc3[n+](c4ccccc4)c2cc1N
IUPAC Name 3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride
Molecular Weight (g/mol) 350.84
Formula Weight 350.1298
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.