missing translation for 'onlineSavingsMsg'
Learn More

Safranin T, Solstice

Product Code. 15623170 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15623170 25 g Glass Bottle 25g
15613170 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15623170 Supplier Honeywell-Fluka Supplier No. 3103125G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Redox indicator and for microscopy, S. No.: 967

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 477-73-6
Molecular Formula C20H19ClN4
Molecular Weight (g/mol) 350.85
MDL Number MFCD00011759
InChI Key QRYAEWIQIBAZOJ-UHFFFAOYSA-N
PubChem CID 2723800
SMILES [Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1

Specifications

Packaging Glass Bottle
UN Number NONH for all modes of transport
Beilstein 3924099
Quantity 25 g
IUPAC Name 3,7-dimethyl-10-phenylphenazin-10-ium-2,8-diamine;chloride
Formula Weight 350.84g/mol
Chemical Name or Material Safranin T
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.