missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SafTest™ Dispenser Set

Product Code. 15732422 Shop All MP Biomedicals Products
Change view
Click to view available options
For Use With (Application):
Alkenal Testing
Free Fatty Acid Testing
Malonaldehyde Testing
Percent Fat Testing
Peroxide Testing
Total Glycerin Testing
Unit Size:
1 set
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) unitSize
15732422 Alkenal Testing 1 set
15742422 Free Fatty Acid Testing 1 set
15772422 Peroxide Testing 1 set
15762422 Percent Fat Testing 1 set
15782422 Total Glycerin Testing 1 set
15752422 Malonaldehyde Testing 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15732422 Supplier MP Biomedicals™ Supplier No. 07DSAK2000
  • ONLINE EXCLUSIVE PRICE
    3130.00 €
    2967.30 € / 1 sets
    Save 162.70 €
    5% Off
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

One dispenser for each reagent (A, B, and preparation Reagent) and one reagent holding rack

Dispenser Set is an accessory for SafTest™ test kits.

  • Dispenser set for each chosen endpoint
  • Allows for accurate testing, every time
  • For use with SafTest™ kits
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Alkenal Testing
Storage Requirements Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.