missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Saikosaponin D

Product Code. 16449036
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449036 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16449036 Supplier MedChemExpress Supplier No. HYN0250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Saikosaponin D is a triterpene saponin isolated from Bupleurum, with anti-inflammatory, anti-bacterial, anti-tumor, and anti-allergic activities; Saikosaponin D inhibits selectin, STAT3 and NF-kB and activates estrogen receptor-β.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 780.98 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 20874-52-6
Product Type Natural Products
Purity 98.0%
Solubility DMSO : 50 mg/mL (64.02 mM∣Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
Target STAT∣NF-κB∣Estrogen Receptor/ERR∣Bacterial
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.