missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint Gobain Life Sciences Chemware™ Cylinder
Graduated Chemware™ Cylinder made from chemically inert fluoropolymers.
Brand: Saint Gobain Life Sciences D1069602
Code : 03
Additional Details : Weight : 1.00000kg
Description
- These graduated cylinders offer all the advantages of non-stick inert fluoropolymers.
- PFA is temperature resistant from -320°F (-196°C) to +500°F (+260°C) and has the lowest metal ion content.
- As a result, Chemware™ cylinders are excellent for trace metal analysis.
Specifications
PFA | |
0.5 mL | |
1.69 oz. | |
Non-sterile |