missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Chemware™ PTFE Solid Balls

Minimize “bumping” in boiling

Brand:  Saint-Gobain

Code : 03

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 10049589

  • 92.45 € / Pack of 100
Estimated Shipment: 31-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • For column packing and as mixing beads
  • Inert to most liquids and vapors
  • Usable to 220°C (428°F)

Specifications

Specifications

PTFE
9.5 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers